missing translation for 'onlineSavingsMsg'
Learn More

Cadmium Standard in Base Oil 75, 1000μg/g, SPEX CertiPrep™

Product Code. 15648325
Click to view available options
Quantity:
50 g
Packaging:
Triple-leached LDPE bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

AnalyteConcentration
Cd1000μg/g

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

CAS 7440-43-9, 8042-47-5
Packaging Triple-leached LDPE bottle
CAS Min % 0.1, 99.9
Quantity 50 g
CAS Max % 0.1, 99.9
Certificate of Analysis With Certificate of Analysis.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Molecular Formula Cd
MDL Number MFCD00010914
InChI Key BDOSMKKIYDKNTQ-UHFFFAOYSA-N
SMILES [Cd]
IUPAC Name cadmium
Molecular Weight (g/mol) 112.41
PubChem CID 23973
ChEBI CHEBI:22977
Grade Oil/Plasma
Concentration or Composition (by Analyte or Components) Matrix Concentration Cat. No.
Base Oil 1000μg/g ORG-CD8-2Z
Base Oil 5000μg/g ORG-CD8-4Z
Solvent or Matrix Base Oil 75
Chemical Name or Material Cadmium Oil Standard
Show More Show Less
compliance-icons
Product Identifier
  • 1000 ug/g Cadmium in Base Oil 75
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
  • Reproductive toxicity Category 2
Hazard Statement
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : cadmium

Product intended for laboratory use only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'