missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Buffer Solution, pH 9.21, METTLER TOLEDO™
Shop All Mettler Toledo Products
Click to view available options
Quantity:
250 mL
1000 mL
6 x 250 mL
Specifications
Specifications
Color | Blue |
pH | 9.21 |
Physical Form | Liquid |
Quantity | 1000 mL |
Solution Type | pH Buffer |
Odor | None |
Packaging | 1000mL |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Buffer Solution, pH 9.21, METTLER TOLEDO™ >
Spot an opportunity for improvement?Share a Content Correction