missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 8.0 (20°C), Buffer solution pH 8.0 (20°C), Honeywell Fluka™

Product Code. 15684000
Click to view available options
Quantity:
1 L

Borax / hydrochloric acid, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Specifications

CAS 1336-21-6
pH 8.0
Quantity 1 L
Molecular Formula H5NO
MDL Number MFCD00066650
UN Number NONH for all modes of transport
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
SMILES N.O
IUPAC Name azanium;hydroxide
Molecular Weight (g/mol) 35.05
PubChem CID 14923
ChEBI CHEBI:18219
Chemical Name or Material Buffer solution
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'