missing translation for 'onlineSavingsMsg'
Learn More

Buffer solution pH 4.0 (20°C), With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST, Honeywell Fluka™

Product Code. 15684020
Click to view available options
Quantity:
100 mL
500 mL
1 L
5 L
10 L
6 x 1 L
This item is not returnable. View return policy
This item is not returnable. View return policy

With fungicide, citric acid / sodium hydroxide / sodium chloride solution, traceable to SRM from NIST

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1336-21-6
Molecular Formula H5NO
Molecular Weight (g/mol) 35.05
MDL Number MFCD00066650
InChI Key VHUUQVKOLVNVRT-UHFFFAOYSA-N
PubChem CID 14923
ChEBI CHEBI:18219
IUPAC Name azanium;hydroxide
SMILES N.O

Specifications

pH 4.0
Quantity 10 L
UN Number NONH for all modes of transport
Chemical Name or Material Buffer solution
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'