missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BUCHI Flask Handler for 20L evaporating flasks
For Rotavapor R-220 models and other 20 L flasks
Brand: BUCHI 041400
This item is not returnable.
View return policy
Description
- Allows for easy handling and mounting
- Compatible with 20L flasks
Specifications
Flask Handler | |
20L flask |