missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTX™ Tweezertrode™ Electrode
Tweezertrode™ Electrode is a reusable, tweezer-style, in vivo electrode suitable for drug or gene delivery in animals.
£1245.00 - £2310.00
Specifications
Product Line | BTX Electroporation |
---|---|
Product Type | Tweezer Style Electrode |
Electrode Shape | Circular |
Quantity | 1 Ea. |
Tip Type | Tweezer Style |
Product Code | Brand | Diameter (Metric) | Material | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Diameter (Metric) | Material | Price | Quantity & Availability | ||||
15487320
|
BTX™
450486 |
1 mm | Platinum |
£1500.00
Each |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15497320
|
BTX™
450487 |
3 mm | Platinum |
£2050.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15417330
|
BTX™
450489 |
5 mm | Platinum |
£1805.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15407330
|
BTX™
450488 |
7 mm | Platinum |
£2310.00
Each |
Estimated Shipment: 21-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15477260
|
BTX™
450118 |
7 mm | Stainless Steel |
£1245.00
Each |
Estimated Shipment: 26-09-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
15487260
|
BTX™
450119 |
10 mm | Stainless Steel |
£1245.00
Each |
Estimated Shipment: 27-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
- Consist of a standard 11.5 cm tweezer modified with disk electrodes of stainless steel
- Compatible with BTX Gemini X2 and ECM-2001 Electroporation Systems
- Gap between electrode disks may be adjusted from 1 mm to more than 2 cm
- Can be easily cleaned
Order Info
Tweerzertrode kits (Cat. No. BTX450486) and additional cable (BTX524) also available separately.
Specifications
BTX Electroporation | |
Circular | |
Tweezer Style | |
Non-sterile |
Tweezer Style Electrode | |
1 Ea. | |
11.5 cm | |
Square Wave |
Intended for Research use only. Not for use in diagnostic or clinical procedures.