Learn More
Thermo Scientific™ BT-549 Control Lysate (1mg/mL)
Thermo Scientific™ BT-549 Control Lysate is a stimulated cell lysate that has been validated for the expression levels and phosphorylation of key targets in the TP53 signaling pathway.
Brand: Thermo Scientific™ A37962
Additional Details : Weight : 0.01000kg
Description
- Validated for the expression of TP53 and phosphorylated TP53 by western blotting and mass spectrometry analysis
- Optimized - stimulation and lysis conditions optimized for detection of TP53 signaling targets
- Convenient - lysate is ready-to-use for immunoprecipitation, immunoprecipitation to mass spectrometry, and western blotting applications
- Stable for two years at -80°C
Specifications
Mass Spectrometry | |
Dry Ice | |
Liquid | |
0.5 mg (1 mg/mL) | |
Protein |
Mass Spectrometer | |
Sample Enrichment | |
Thermo Scientific™ | |
Control Lysate | |
Cell Lysate |
For Research Use Only. Not for use in diagnostic procedures.