missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals Bromocresol Purple, pure, indicator grade

Product Code. 15760809
Click to view available options
Quantity:
5 g
25 g
140 g
1 kg
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Indicator
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 115-40-2
Molecular Formula C21H16Br2O5S
Molecular Weight (g/mol) 540.22
MDL Number MFCD00011681
InChI Key ABIUHPWEYMSGSR-UHFFFAOYSA-N
Synonym Bromcresol Purple, BCP, 5', 5''-Dibromo-o-cresolsulfonephthalein
PubChem CID 8273
ChEBI CHEBI:86154
SMILES CC1=CC(=CC(Br)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C)=C(O)C(Br)=C1

Specifications

Absorbance (20 mg/L in methanol) Complies, (20 mg/L in methanol) Complies at 0nm
Melting Point 245.0°C
Solubility Information Solubility in water: practically insoluble. Other solubilities: 75g/L in ethanol (20°C)
Infrared Spectrum Authentic
IUPAC Name 2-bromo-4-[3-(3-bromo-4-hydroxy-5-methylphenyl)-1,1-dioxo-2,1$l^{6}-benzoxathiol-3-yl]-6-methylphenol
Formula Weight 540.21
Grade Pure
Packaging Glass bottle
Visual Transition Interval from pH 5.2 (yellow) to pH 6.8 (purple)
Color Pink or Salmon
Quantity 140 g
Physical Form Powder
Chemical Name or Material Bromocresol Purple
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'