missing translation for 'onlineSavingsMsg'
Learn More
Bromate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ Product Code.: 12992085

Bromate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™
BIOPHARMA

Product Code. 12992085
250 mL, Bottle
Click to view available options
Quantity:
100 mL
250 mL
500 mL
Packaging:
Bottle
This item is not returnable. View return policy

Product Code. 12992085

Brand: Fisher Chemical A 007 250

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.

Chemical Identifiers

Molecular Formula BrO3-

Specifications

Packaging Bottle
Quantity 250 mL
Physical Form Liquid
Concentration 1 g/L
Type Single-component Standard Solution
Solvent or Matrix H2O
Chemical Name or Material Bromate
compliance-icons
Product Identifier
  • Single-component Standard Solution for Ion chromatography. Bromate 1000mg/l in water (A 007)
Signal Word
  • Danger
Hazard Category
  • Carcinogenicity Category 1B
Hazard Statement
  • H350-May cause cancer.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P202-Do not handle until all safety precautions have been read and understood.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Bromate solution for Ion Chromatography 1g/L H2O matrix, Fisher chemical™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.