Learn More
JG Finneran Associates™ Flat Bottom Inserts
Designed for use in 2mL standard opening crimp top and screw thread vials. JG Finneran™ Flat Bottom Inserts are an economical alternative to the conical limited volume inserts.
Brand:  JG Finneran Associates™ 4035-630
Code : 17
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12307029
  • £219.82 / Pack of 1000
Estimated Shipment: 16-07-2024
to see stock.
Add to basket
Description
Description
  • Flat Bottom Glass and Polypropylene Inserts
  • With ID ring at the top of the insert for easy open-end identification
  • Available salinized or pre-cleaned
Specifications
Specifications
Glass
350 μL
Flat
1000 Pk.