Learn More
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate
For the safe and secure closure of flasks, bottles, tubing and other laboratory apparatus. Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate are manufactured from durable DURAN™ borosilicate glass.
£30.30 - £60.07
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Standard Taper Joint | Price | Quantity & Availability | ||||||
11728034
|
Lenz Laborglasinstrumente™
01400312 |
NS 12/21 |
£30.30
Pack of 10 |
Estimated Shipment: 28-05-2025
Log in to see stock. |
||||||
11738034
|
Lenz Laborglasinstrumente™
01400314 |
NS 14/23 |
£34.68
Pack of 10 |
Estimated Shipment: 12-05-2025
Log in to see stock. |
||||||
11718034
|
Lenz Laborglasinstrumente™
01400310 |
NS 10/19 |
£32.55
Pack of 10 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11748034
|
Lenz Laborglasinstrumente™
01400319 |
NS 19/26 |
£40.78
Pack of 10 |
Estimated Shipment: 20-05-2025
Log in to see stock. |
||||||
11758034
|
Lenz Laborglasinstrumente™
01400324 |
NS 24/29 |
£51.48
Pack of 10 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
11876164
|
Lenz Laborglasinstrumente™
01400334 |
NS 34/35 |
£59.60
Pack of 5 |
Estimated Shipment: 06-05-2025
Log in to see stock. |
||||||
11768034
|
Lenz Laborglasinstrumente™
01400329 |
Thread Size: 29/32 Cone NS |
£60.07
Pack of 10 |
Estimated Shipment: 16-05-2025
Log in to see stock. |
||||||
Description
- Available in a range of cone sizes
- Corrosion-free and resistant to thermal shock
Specifications
Borosilicate Glass | |
Hollow Stopper |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Lenz Laborglasinstrumente™ Flat Bottom Hollow Stoppers, Hexagonal Plate