missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis(tri-n-butyltin) oxide, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Bis(tri-n-butyltin) oxide is employed in the synthesis of α,β-unsaturated methyl ketones, isoxazoles.
Solubility
Soluble in water (0.071 g/L)
Notes
Store at room temperature. Incompatible with oxidizing agents. Ensure adequate ventilation.
Specifications
Specifications
Melting Point | -45°C |
Density | 1.172 |
Boiling Point | 179°C to 180°C (2 mmHg) |
Flash Point | 168°C (334°F) |
Odor | Irritating |
Linear Formula | [(CH3CH2CH2CH2)3Sn]2O |
Refractive Index | 1.4865 |
Quantity | 2500 g |
UN Number | UN2788 |
Beilstein | 745057 |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'