missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Bis[2-(di-tert-butylphosphino)ethyl]amine, 10% w/w soln. in THF, Thermo Scientific Chemicals
Click to view available options
Quantity:
1 g
5 g
25 g
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
This compound is involved in the transition metal-catalyzed addition of amines to acrylic acid derivatives.
Solubility
Insoluble in water.
Notes
Air Sensitive. Incompatible with oxidizing agents.
Specifications
Density | 0.911 |
Odor | Unpleasant |
Linear Formula | ([(CH3)3C]2PCH2CH2)2NH |
Refractive Index | 1.418 |
Quantity | 1 g |
UN Number | UN1993 |
Sensitivity | Air Sensitive |
Solubility Information | Insoluble in water. |
Formula Weight | 361.53 |
Concentration or Composition (by Analyte or Components) | 10% w/w soln. in THF |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'