missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta - Nicotinamide Adenine Dinucleotide Phosphate Reduced Tetrasodium Salt 98% MP Biomedicals
Click to view available options
Quantity:
25 mg
50 mg
100 mg
250 mg
500 mg
1 g
Specifications
Assay Percent Range | 98% |
Quantity | 25 mg |
SMILES | [Na+].[Na+].[Na+].[Na+].NC(=O)C1=CN(C=CC1)[C@@H]1O[C@H](COP([O-])(=O)OP([O-])(=O)OC[C@H]2O[C@H]([C@H](OP([O-])([O-])=O)[C@@H]2O)N2C=NC3=C(N)N=CN=C23)[C@@H](O)[C@H]1O |
IUPAC Name | {[(2R,3R,4R,5R)-2-(6-amino-9H-purin-9-yl)-5-[({[({[(2R,3S,4R,5R)-5-(3-carbamoyl-1,4-dihydropyridin-1-yl)-3,4-dihydroxyoxolan-2-yl]methoxy}(hydroxy)phosphoryl)oxy](hydroxy)phosphoryl}oxy)methyl]-4-hydroxyoxolan-3-yl]oxy}phosphonic acid |
Formula Weight | 833.4 |
Percent Purity | 98% |
Physical Form | Powder |
Chemical Name or Material | Beta - Nicotinamide Adenine Dinucleotide Phosphate, Reduced Tetrasodium Salt |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'