missing translation for 'onlineSavingsMsg'
Learn More

beta-D-Ribofuranose 1,2,3,5-tetraacetate, 98+%, Thermo Scientific Chemicals

£26.33 - £215.00

Chemical Identifiers

CAS 13035-61-5
Molecular Formula C13H18O9
Molecular Weight (g/mol) 318.278
MDL Number MFCD00005358
InChI Key IHNHAHWGVLXCCI-FDYHWXHSSA-N
Synonym beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
PubChem CID 83064
IUPAC Name [(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
SMILES CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11475877
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24426.06
5 g
£26.33
5g
Please to purchase this item. Need a web account? Register with us today!
11485877
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24426.14
25 g
£76.18
25g
Please to purchase this item. Need a web account? Register with us today!
11495877
View Documents Promotion Details
Thermo Scientific Alfa Aesar
B24426.22
100 g
£215.00
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

13035-61-5
318.278
IHNHAHWGVLXCCI-FDYHWXHSSA-N
83064
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
C13H18O9
MFCD00005358
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate

Specifications

13035-61-5
C13H18O9
94078
IHNHAHWGVLXCCI-FDYHWXHSSA-N
CC(=O)OCC1C(C(C(O1)OC(=O)C)OC(=O)C)OC(=O)C
318.278
318.28
beta-D-Ribofuranose 1,2,3,5-tetraacetate
80°C to 83°C
MFCD00005358
beta-d-ribofuranose 1,2,3,5-tetraacetate, tetra-o-acetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-beta-d-ribofuranose, tetraacetylribose, 1,2,3,5-tetraacetyl-.-d-ribofuranose, tetra-o-acetyl-b-d-ribofuranose, 1,2,3,5-tetraacetyl-beta-d-ribofuranose, 1,2,3,5-tetra-o-acetyl-b-d-ribofuranose, .beta.-d-ribofuranose, tetraacetate, ribofuranose, tetraacetate, .beta.-d
−13±0.5° (c=2.4 in Chloroform)
[(2R,3R,4R,5S)-3,4,5-triacetyloxyoxolan-2-yl]methyl acetate
83064
≥98%
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 235-898-5

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only