missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate, ≥95%, Thermo Scientific™
£69.00 - £229.00
Chemical Identifiers
CAS | 135829-04-8 |
---|---|
Molecular Formula | C17H17NO3 |
Molecular Weight (g/mol) | 283.327 |
MDL Number | MFCD02682013 |
InChI Key | QSMOQTIGILELKY-UHFFFAOYSA-N |
Synonym | benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline |
PubChem CID | 2776402 |
IUPAC Name | benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate |
SMILES | C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10722673
|
Thermo Scientific Maybridge
CC13409DA |
1 g |
£69.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10690656
|
Thermo Scientific Maybridge
CC13409DE |
5 g |
£229.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
135829-04-8 | |
283.327 | |
QSMOQTIGILELKY-UHFFFAOYSA-N | |
2776402 | |
C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO |
C17H17NO3 | |
MFCD02682013 | |
benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline | |
benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate |
Specifications
135829-04-8 | |
100.0 | |
Amber Glass Bottle | |
MFCD02682013 | |
QSMOQTIGILELKY-UHFFFAOYSA-N | |
benzyl 2-(hydroxymethyl)-2,3-dihydroindole-1-carboxylate | |
2776402 | |
95% |
94.5 | |
95% | |
C17H17NO3 | |
benzyl 2-hydroxymethyl-1-indolinecarboxylate, benzyl 2-hydroxymethyl indoline-1-carboxylate, benzyl 2-hydroxymethyl-2,3-dihydroindole-1-carboxylate, 1h-indole-1-carboxylicacid, 2,3-dihydro-2-hydroxymethyl-, phenylmethyl ester, acmc-20c3wg, 1-benzyloxycarbonylindoline-2-methanol, +/-1-benzyloxycarbonyl-2-hydroxymethylindoline | |
C1C(N(C2=CC=CC=C21)C(=O)OCC3=CC=CC=C3)CO | |
283.327 | |
283.33 | |
Benzyl 2-(hydroxymethyl)-1-indolinecarboxylate |