Learn More
Bel-Art™ SP Scienceware™ Polystop™ Dropper Bottles
Prevent chipping of the glass dropper upon reinsertion into the dropper bottle
£22.18 - £22.49
Specifications
Color Clear
Material Glass
Closure Material Polypropylene
View More Specs
Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10389547
View Documents Promotion Details
Bel-Art™
F11604-0030
30 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10521958
View Documents Promotion Details
Bel-Art™
F11604-0060
60 mL This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description
Description
  • The dropper easily pulls free, while the stopper remains snugly in the neck of the bottle, thus providing a safe return space for the dropper
  • Polypropylene stopper
  • Clear glass bottle has a ground T-neck, and the dropper has a soft vinyl bulb
  • Disassembles for easy cleaning
Specifications
Specifications
Clear
Glass
Polypropylene