Learn More
Bel-Art™ SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars in Colors
Registered ISO 9001:2008. SP Scienceware™ Octagon Spinbar™ Magnetic Stir Bars in Colors are the most commonly used shape.
£201.00
Description
- PTFE-coated
- With pivot ring
- Assorted sizes and colors
Specifications
Octagon |