missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Catalase Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Brand: BD 261203
This item is not returnable.
View return policy
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Specifications
Reagent/Stain Dropper | |
CE | |
Dropper | |
15° to 30°C |
50 Pack | |
One day's worth of stain | |
Used for determining the presence of catalase produced by bacteria |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Catalase Test
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P321-Specific treatment (see on this label).
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P330-Rinse mouth.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrogen peroxide solution