missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD FoxP3 Staining Kit
High performance reagent kit for detection of human FoxP3 positive regulatory T cells
£1620.00
Includes: PE/Alexa Fluor™ 647/ Alexa Fluor 488, FoxP3, CD4, CD25, Buffer set
Product Code | Brand | Conjugate | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Conjugate | Price | Quantity & Availability | |||||
15829958
|
BD
560131 |
Alexa Fluor 488 |
£1620.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15839958
|
BD
560132 |
Alexa Fluor 647 |
£1620.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
15849958
|
BD
560133 |
PE |
£1620.00
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||||
Description
- Complementing antibody conjugates buffer system fixes and permeabilizes cells in easy, simple steps
- Components are sufficient for 100 tests
- Clone: M-A251, RPA-T4, 259D/C7
- Isotype: mouse IgG1, IgG1, κ, IgG1, κ
Flow Cytometry
Research use only