missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Mitis Salivarius Agar
Use with Chapman Tellurite Solution 1% for selective isolation of S. mitis, S. salivarius and enterococci
Brand: BD 229810
Additional Details : Weight : 0.80000kg
Specifications
CE | |
Dehydrated | |
Agar | |
Used with Tellurite solution 1% in isolating Streptococcus mitis, S. salivarius and enterococci, particularly from grossly contaminated specimen | |
500 g |
Refrigerate | |
Mitis Salivarius Agar | |
Pancreatic digest of Casein 6g/L, Proteose Peptone 9g/L, Proteose Peptone 5g/L, Dextrose 1g/L, Saccharose 50g/L, Dipotassium Phosphate 4g/L, Trypan blue 75mg/L, Crystal Violet 0.8mg/L, Agar 15g/L | |
Bottle |