missing translation for 'onlineSavingsMsg'
Learn More

BD Difco™ Dehydrated Culture Media: Malt Extract Agar

For isolation, detection and enumeration of yeasts and molds

Brand:  BD 211220

Additional Details : CAS Number : 9004-53-9 Weight : 0.80000kg

Product Code. 11778113

  • £179.00 / 500g
Estimated Shipment: 25-07-2024
to see stock.
Add to basket

Description

Description

Order Info

See our web site or the Fisher Scientific™ HealthCare catalog for product information.

Certifications

ISO 9001:2008, BAM, COMPF

Specifications

Specifications

ISO 9001:2008, BAM, COMPF
Dehydrated
Agar
Isolating, cultivating and enumerating Yeasts and mold
500 g
Refrigerate
Malt Extract Agar
Maltose 12.75g/L, Dextrin 2.75g/L, Glycerol 2.35g/L, Peptone 0.78g/L, Agar 15g/L
Bottle
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers