missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: ISP Medium 2
International Streptomyces Project Yeast Malt Extract Agar
Brand: BD 277010
Additional Details : Weight : 0.53100kg
Specifications
CE | |
Dehydrated | |
Agar | |
For characterizing Streptomyces species according to the International Streptomyces project | |
500 g |
Refrigerate | |
ISP-Yeast Malt Agar (Medium 2) | |
Yeast extract 4g/L, Malt extract 10g/L, Dextrose 4g/L, Agar 20g/L | |
Bottle |