Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Rosolic Acid
Use with mFC Broth Base and mFC Agar for detection and enumeration of fecal coliforms
Brand: BD 232281
Additional Details : CAS Number : 603-45-2 Weight : 0.80000kg
Specifications
ISO/DIS, 11014 | |
Rosolic Acid | |
Detection and enumeration of fecal Coliforms, m-FC broth base and m-FC Agar | |
1 g |
<30°C | |
Crystalline Powder | |
Vial |
Safety and Handling
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with all local/regional/national/internationalregulations.
Supplemental information
- MIXTURE LIST-Contains : rosolic acid