missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Acridine Orange Stain
For detecting microorganisms in direct smears by fluorescence
Brand: BD 212536
This item is not returnable.
View return policy
Description
- Especially beneficial in screening normally sterile specimens where few organisms may be present
- Bacteria stain bright orange; human cells, tissue debris and proteinaceous material stain pale green to yellow

Specifications
Acridine Orange Stain | |
CE | |
Acridine Orange |
250 mL | |
Recommended for use in the fluorescent microscopic detection of microorganisms in direct smears prepared from clinical and non clinical materials |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P271-Use only outdoors or in a well-ventilated area.
- P321-Specific treatment (see on this label).
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P405-Store locked up.
- P403+P233-Store in a well-ventilated place. Keep container tightly closed.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : hydrochloric acid
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
BD Acridine Orange Stain > 1 x 250mL; 1 Ea.
Spot an opportunity for improvement?Share a Content Correction