Learn More
Julabo™ Bath Adapters
Brand:  Julabo™ 8890110
 View more versions of this product
Product Code. 15244293
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Magnum 91, PRESTO™ A30, PRESTO™ A40, PRESTO™ A80, PRESTO™ W40 temperature control/process system
Adapter
M24 x 1/2 male to M24 x 1/2 male