missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium molybdenum oxide, 99.9% (metals basis excluding Sr), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Barium molybdenum oxide is used in electronic and optical equipment, pigment in paints and other protective coatings. It is also used as enamel of the adhesive products for iron's adhesion strong.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∼1600°C |
Color | White |
Physical Form | Powder |
Quantity | 25 g |
UN Number | UN1564 |
Solubility Information | Slightly soluble in water and acids |
Formula Weight | 297.28 |
Percent Purity | 99.9% |
Odor | Odorless |
Density | 4.98g/mL |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'