missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Barium fluoride, 99.9% (metals basis), Thermo Scientific Chemicals
Click to view available options
Quantity:
25 g
100 g
Description
Barium fluoride is used as a material to make optical components such as lenses, in windows for infrared spectroscopy, in particular in the field of fuel oil analysis, used as a preopacifying agent and in enamel and glazing frits production. It is also used in the production of welding agents, metallurgy, as a molten bath for refining aluminum.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Boiling Point | 2260°C |
Melting Point | 1353°C |
Color | Colorless |
Physical Form | Fused Lump |
Quantity | 25 g |
Assay Percent Range | 99.99% (Metals basis) |
Solubility Information | Soluble in methanol,ethanol,water 1.2g/L. |
IUPAC Name | barium(2+) difluoride |
Formula Weight | 175.36 |
Odor | Odorless |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'