missing translation for 'onlineSavingsMsg'
Learn More

Barium chloride dihydrate, Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur., ≥99%, Honeywell Fluka™

Product Code. 15653250
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
5 kg
4 x 5 kg
50 kg
6 x 1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Puriss. p.a. plus, ACS Reagent, Reag. ISO, Reag. Ph. Eur.,≥99%

  • Exceeds standard ACS specifications with additional trace metals analysis
  • Products with a guarantee certificate and/or suitable for the stated analytical application.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10326-27-9
Molecular Formula BaCl2H4O2
Molecular Weight (g/mol) 244.26
MDL Number MFCD00149154
InChI Key PWHCIQQGOQTFAE-UHFFFAOYSA-L
Synonym barium chloride dihydrate, bacl2.2h2o, barium 2+ dihydrate dichloride, barium 2+ dichloride dihydrate, acmc-20alrc, dsstox_cid_131, barium chloride, acs, dsstox_rid_75389, dsstox_gsid_20131, ksc174q7d
PubChem CID 5284346
ChEBI CHEBI:86153
IUPAC Name barium(2+);dichloride;dihydrate
SMILES O.O.[Cl-].[Cl-].[Ba++]

Specifications

Quantity 2.5 kg
UN Number 1564
Formula Weight 244.26g/mol
Percent Purity ≥99%
Grade ACS/puriss. p.a. plus
Packaging Plastic Bottle
Chemical Name or Material Barium Chloride Dihydrate, Reag. Ph. Eur. reag. ISO
compliance-icons
Product Identifier
  • Barium chloride dihydrate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Acute toxicity Category 4
Hazard Statement
  • H301-Toxic if swallowed.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'