missing translation for 'onlineSavingsMsg'
Learn More

Azpack™ Cargille™ Immersion Oil

Choose between three types for normal or fluorescence microscopy. Azpack™ Cargille™ Immersion Oil is supplied with a listing of optical and physical properties.

Brand:  Azpack™ 12423170

22.52 GBP valid until 2024-04-30
Use promo code "21637" to get your promotional price.



Code : 04

Additional Details : Weight : 0.06840kg


 View more versions of this product

Product Code. 12423170

  • £104.00 / Each
Estimated Shipment: 18-06-2024
to see stock.
Add to basket

Description

Description

  • Supplied with specific optical and physical properties listing
  • For use with normal light or fluorescence microscopy
Specifications

Specifications

Immersion Oil Type B
1250 cSt
Normal light or fluorescence microscopy
30 mL
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • 30 ML OIL IMMERSION CARGILLE B FOR EDUCATIONAL

Signal Word
  • Warning

Hazard Category
  • Skin corrosion/irritation Category 2

Hazard Statement
  • H315-Causes skin irritation.

Precautionary Statement
  • P264-Wash thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.

Supplemental information
  • MIXTURE LIST-Contains : Trade Secret

SDS
Documents

Documents

Certificates
Special Offers

Special Offers