Learn More
Azpack™ Cargille™ Immersion Oil
Choose between three types for normal or fluorescence microscopy. Azpack™ Cargille™ Immersion Oil is supplied with a listing of optical and physical properties.
Brand: Azpack™ 11338113
Code : 03
Additional Details : Weight : 0.20340kg
Description
- Supplied with specific optical and physical properties listing
- For use with normal light or fluorescence microscopy
Specifications
Immersion Oil Type B | |
1250 cSt | |
Normal light or fluorescence microscopy | |
120 mL |
Safety and Handling
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
Precautionary Statement
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.