missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azlon™ High Temperature Polypropylene Autoclave Bags
Autoclave waste in this autoclave bag for biohazardous material. The Azlon™high temperature polypropylene autoclave bag has printed instructions and can be autoclaved at temperatures up to 135 degrees C.
Brand: Azlon BWY250
Code : 02
Additional Details : Weight : 1.93000kg
Description
- Suitable for autoclaving biohazardous waste
- High Temperature suitability
- Strong side weld construction (50μm)
- Printed with instructions for use and biohazard symbol
Specifications
Autoclavable | |
Polypropylene | |
660 mm |
Clear | |
302 mm |