Learn More
Azlon™ Multi-lingual Round Integral Wash Bottle
Product Code. 15828477
Click to view available options
Chemical Label:
Acetone
Distilled Water
Ethanol
Isopropanol
Methanol
Azlon™ Multi-lingual Round Integral Wash Bottle is molded with an integral spout to reduce contamination caused by the inner tube touching the benchtop during re-filling.
  • Multilingual
  • Wide neck wash bottle
  • Indelibly printed in English, French, German and Spanish
  • Choose from five solvent labels all including the new Globally Harmonised Symbols, NFPA diamond, risk phrases and safety information
  • Fitted with color coded one-piece PP closure that maintains it's shape in use
  • Designed with integral spout to prevent contamination
TRUSTED_SUSTAINABILITY
Specifications
Capacity (Metric) 500 mL
Includes Integral spout
Closure Color Orange
Closure Included Yes
Mouth Wide
Product Type Wash Bottle
Chemical Label Ethanol
Closure Material Polypropylene