Learn More
Deltalab™ Autoclave Resistant Bags
Wrap autoclaveable items in Deltalab™Autoclave Resistant Bags when working with laboratory items requiring sterilization.
POA
Specifications
Autoclavable Autoclavable
View More Specs
Description
Description
  • Printed with biohazard graphic symbol and use instructions in five languages
  • ags placed in autoclave at 121ºC. should not be sealed hermetically
  • Minimum order quantity: 50 pieces
Specifications
Specifications
Autoclavable