Learn More
Labcaire™ AU-1210-AC-R Fume Cabinet Filter
Brand: Labcaire™ AU-1210
Code : 04
Additional Details : Weight : 12.00000kg
Specifications
AC-R | |
12 Kg | |
AU-1210-AC-R |
Aura Model 250/550 | |
Fume Cabinet Filter | |
Main For Radioactive Compounds - Iodine 125 |