missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nabertherm Ashing Furnace, LVT Series
Designed for ashing in the laboratory. A special air intake and exhaust system allows air exchange of more than 6 times per minute. Incoming air is preheated to ensure a good temperature uniformity.
£3177.30 - £3728.10
Product Code | Brand | Capacity (Metric) | Temperature Control | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Temperature Control | Price | Quantity & Availability | ||||||
15589572
|
Nabertherm
LV052H1RN1 |
5 L | B410 | N/A | N/A | N/A | |||||
15908370
|
Nabertherm
LV052H1TN1 |
5 L | C450 | N/A | N/A | N/A | |||||
15519582
|
Nabertherm
LV092H1RN1 |
9 L | B410 | N/A | N/A | N/A | |||||
Description
- Maximum Temperature: 1100 °C
- Heating from two sides
- Ceramic heating plates with integral heating element which is safeguarded against fumes and splashing, and easy to replace
- Air exchange of more than 6 times per minute
- Good temperature uniformity due to preheating of incoming air
- Only fiber materials are used which are not classified as carcinogenic according to TRGS 905, class 1 or 2
- Housing made of sheets of textured stainless steel
- Dual shell housing for low external temperatures and stability
- Lift door with hot surface facing away from the operator
- Solid state relays provide for lownoise operation
- Defined application within the constraints of the operating instructions
- NTLog Basic for Nabertherm controller: recording of process data with USB-flash drive
Specifications
1100°C |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nabertherm Ashing Furnace, LVT Series