Learn More
Applied Biosystems™ GeneAmp™ dNTP Blend (2.5 mM ea)
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.
Brand: Applied Biosystems™ 4303442
Code : NEW
Additional Details : Weight : 0.01000kg
Description
GeneAmp dNTPs are deoxynucleotide triphosphates dissolved in glass-distilled water and titrated to pH 7.0 with NaOH. Each vial consists of 1 mL of 2.5 mM each of dATP, dCTP, dGTP, and dTTP, yielding a 10 mM blend of dNTPs.
Key advantages of GeneAmp dNTPs
• High purity dNTPs are ideal for use in PCR reactions or other applications
• Guaranteed for PCR performance when used with GeneAmp PCR core reagents and GeneAmp instrument systems
• Premixed dNTP blends are available in various concentrations, offering ease of use
Specifications
GeneAmp dNTP Blend, 2.5 mM (12 x 1 mL) Store in a constant temperature freezer at –20°C. |
|
GeneAmp™ | |
Dry Ice | |
10 mM |
Liquid | |
12 x 1 mL | |
Standard PCR |
For Research Use Only. Not for use in diagnostic procedures.