Learn More
Antimony(III) sulfide, 99.999% (metals basis), Thermo Scientific Chemicals
£25.12 - £548.73
Chemical Identifiers
CAS | 1345-04-6 |
---|---|
Molecular Formula | S3Sb2 |
Molecular Weight (g/mol) | 339.70 |
MDL Number | MFCD00011217 |
InChI Key | NVWBARWTDVQPJD-UHFFFAOYSA-N |
IUPAC Name | diantimony(3+) trisulfanediide |
SMILES | [S--].[S--].[S--].[Sb+3].[Sb+3] |
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | ||||||
11381788
|
Thermo Scientific Alfa Aesar
011683.04 |
2 g | N/A | N/A | N/A | |||||
11371788
|
Thermo Scientific Alfa Aesar
011683.09 |
10 g |
£104.52
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
11391788
|
Thermo Scientific Alfa Aesar
011683.18 |
50 g |
£548.73
50g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
It is used in glitter compositions, fountain compositions, flash powder and vulcanizing rubber. It is employed in the production of ruby-colored glass and in plastics, as a fire retardant, in explosives, matches, and fireworks. It can be used to prepare a series of thioantimonous salts and Schlippe's salt.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1345-04-6 | |
339.70 | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
[S--].[S--].[S--].[Sb+3].[Sb+3] |
S3Sb2 | |
MFCD00011217 | |
diantimony(3+) trisulfanediide |
Specifications
550°C | |
Powder | |
MFCD00011217 | |
14,713 | |
[S--].[S--].[S--].[Sb+3].[Sb+3] | |
339.70 | |
99.999% | |
Antimony(III) sulfide |
1345-04-6 | |
S3Sb2 | |
UN1549 | |
NVWBARWTDVQPJD-UHFFFAOYSA-N | |
diantimony(3+) trisulfanediide | |
339.68 | |
(metals basis) |
Safety and Handling
GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Warning
missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
missing translation for 'tsca' : TSCA
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.