Learn More
WNT8A Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA534330
Code : NEW
Additional Details : Weight : 0.01000kg
Description
Percent identity with other species by BLAST analysis: Human, Gorilla, Gibbon, Bat (100%) Monkey, Marmoset, Mouse, Panda, Dog, Horse (94%) Rat, Rabbit (89%) Elephant, Bovine (83%).
WNT8A, part of the WNT gene family, consists of structurally related genes that encode secreted signaling proteins. These proteins have been implicated in oncogenesis and in several developmental processes, including regulation of cell fate and patterning during embryogenesis. This gene is a member of the WNT gene family. Study of its expression in the teratocarcinoma cell line NT2 suggests that it may be implicated in the early process of neuronal differentiation of NT2 cells induced by retinoic acid. This gene is clustered with WNT3, another family member, in the chromosome 17q21 region.Specifications
WNT8A | |
Polyclonal | |
Unconjugated | |
Wnt8a | |
etID309727.14; protein Wnt-8a; protein Wnt-8a ORF1; protein Wnt-8d; Stimulated by retinoic acid gene 11 protein; Stra11; wingless-related MMTV integration site 8A; wingless-related MMTV integration site 8D; wingless-type MMTV integration site family member 8A; wingless-type MMTV integration site family, member 8A; Wnt family member 8A; wnt8; wnt-8; wnt8.1; Wnt8a; Wnt-8A; Wnt8d; Wnt-8D; Wnt8-like protein 1; wu:fa20e02; wu:fe05d07 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7478 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
PBS with 0.1% sodium azide | |
Q9H1J5 | |
Wnt8a | |
Synthetic 18 amino acid peptide from internal region of human WNT8A. | |
50 μg | |
Primary | |
Human | |
Antibody | |
IgG |