Learn More
NSD2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA540725
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: KYNVGDLVWS KVSGYPWWPC MVSADPLLHS YTKLKGQKKS ARQYHVQFFG Sequence homology: Dog: 93%; Guinea Pig: 88%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 76%; Rat: 86%
NSD2 Polyclonal antibody specifically detects NSD2 in Human samples. It is validated for Western Blot.Specifications
NSD2 | |
Polyclonal | |
Unconjugated | |
NSD2 | |
5830445G22Rik; 9430010A17Rik; AW555663; C130020C13Rik; D030027O06Rik; D930023B08Rik; Histone-lysine N-methyltransferase NSD2; histone-lysine N-methyltransferase NSD2; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; IL5 promoter REII region-binding protein; Kiaa1090; LOW QUALITY PROTEIN: histone-lysine N-methyltransferase NSD2; mKIAA1090; MMSET; multiple myeloma SET domain containing protein type III; multiple myeloma SET domain-containing protein; NSD2; nuclear receptor binding SET domain protein 2; nuclear SET domain-containing protein 2; probable histone-lysine N-methyltransferase NSD2; Protein trithorax-5; putative histone-lysine N-methyltransferase NSD2-like protein; REIIBP; RGD1565590; trithorax/ash1-related protein 5; TRX5; WHS; Whsc1; Whsc1l; Wolf-Hirschhorn syndrome candidate 1; Wolf-Hirschhorn syndrome candidate 1 (human); Wolf-Hirschhorn syndrome candidate 1 protein; Wolf-Hirschhorn syndrome candidate 1 protein homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
7468 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
O96028 | |
NSD2 | |
Synthetic peptide directed towards the N-terminal of human WHSC1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |