missing translation for 'onlineSavingsMsg'
Learn More
Learn More
WDHD1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539532
Code : NEW
Additional Details : Weight : 0.00900kg
Description
The protein encoded by this gene contains multiple N-terminal WD40 domains and a C-terminal high mobility group (HMG) box. WD40 domains are found in a variety of eukaryotic proteins and may function as adaptor/regulatory modules in signal transduction, pre-mRNA processing and cytoskeleton assembly. HMG boxes are found in many eukaryotic proteins involved in chromatin assembly, transcription and replication. Alternative splicing results in two transcript variants encoding different isoforms. [provided by RefSeq, Jul 2008].Specifications
WDHD1 | |
Polyclonal | |
Unconjugated | |
WDHD1 | |
AND1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11169 | |
-20°C | |
Liquid |
Western Blot | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O75717 | |
WDHD1 | |
A synthetic peptide derived from the internal region of human WDHD1 | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |