missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SMAD6 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA525263
Code : LD
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse based on sequence homology.
SMAD6 (mothers against decapentaplegic homolog 6) is an antagonist of signaling by TGF-beta (transforming growth factor) type 1 receptor superfamily members; has been shown to inhibit selectively BMP (bone morphogenetic proteins) signaling by competing with the co-SMAD SMAD4 for receptor-activated SMAD1. SMAD6 is an inhibitory SMAD (I-SMAD) or antagonistic SMAD.Specifications
SMAD6 | |
Polyclonal | |
Unconjugated | |
SMAD6 | |
AOVD2; b2b390Clo; HsT17432; MAD homolog 6; mad homolog 7; Madh6; Madh7; Mothers against decapentaplegic homolog 6; mothers against decapentaplegic, drosophila, homolog of, 6; mothers against DPP homolog 6; Msmad6; SMAD; SMAD 6; SMAD family member 6; SMAD SMAD; SMAD, mothers against DPP homolog 6; SMAD6 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4091 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.46 mg/mL | |
PBS with 0.09% sodium azide | |
O43541 | |
SMAD6 | |
KLH conjugated synthetic peptide between 358-386 amino acids from the central region of human SMAD6 | |
400 μL | |
Primary | |
Human | |
Antibody | |
Ig |