missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC28A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA537223
Code : NEW
Additional Details : Weight : 0.00910kg
Description
This antibody detects endogenous protein at a molecular weight of 71 kDa. Purity is >95% by SDS-PAGE.
SLC28A1 Polyclonal antibody specifically detects SLC28A1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
SLC28A1 | |
Polyclonal | |
Unconjugated | |
Slc28a1 | |
CNT 1; CNT1; Concentrative nucleoside transporter 1; HCNT1; Na(+)/nucleoside cotransporter 1; SLC28A1; sodium/nucleoside cotransporter 1; Sodium-coupled nucleoside transporter 1; solute carrier family 28 (concentrative nucleoside transporter), member 1; solute carrier family 28 (sodium-coupled nucleoside transporter), member 1; solute carrier family 28 member 1; solute carrier family 28, member 1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
116642, 434203, 9154 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide | |
O00337, Q62674 | |
Slc28a1 | |
A synthetic peptide corresponding to residues in human SLC28A1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |