Learn More
SLC25A29 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542417
Code : NEW
Additional Details : Weight : 0.00010kg
Description
Peptide sequence: AEGWRVFTRG LASTLLRAFP VNAATFATVT VVLTYARGEE AGPEGEAVPA Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 93%
SLC25A29 Polyclonal antibody specifically detects SLC25A29 in Human samples. It is validated for Western Blot.Specifications
SLC25A29 | |
Polyclonal | |
Unconjugated | |
Slc25a29 | |
C030003J19Rik; C14orf69; CACL; CACT-like; carnitine/acylcarnitine translocase-like; carnitine-acylcarnitine translocase like; hypothetical protein LOC569608; LOC100715169; LOW QUALITY PROTEIN: mitochondrial basic amino acids transporter; mCACL; mitchondrial carnitine/acylcarnitine carrier protein CACL; mitochondrial basic amino acids transporter; Mitochondrial carnitine/acylcarnitine carrier protein CACL; mitochondrial ornithine transporter 3; Ornt3; Slc25a29; solute carrier family 25 (mitochondrial carnitine/acylcarnitine carrier), member 29; solute carrier family 25 (mitochondrial carrier, palmitoylcarnitine transporter), member 29; solute carrier family 25 member 29; solute carrier family 25, member 29; zgc:114094 | |
Rabbit | |
Protein A | |
RUO | |
123096 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
1 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q8N8R3 | |
Slc25a29 | |
Synthetic peptide directed towards the C-terminal of human SLC25A29 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |