Learn More
SIN3B Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA530707
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: HepG2, HCT116, mouse brain. Predicted reactivity: Mouse (92%), Rat (91%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
Acts as a transcriptional repressor. Interacts with MXI1 to repress MYC responsive genes and antagonize MYC oncogenic activities. Interacts with MAD-MAX heterodimers by binding to MAD. The heterodimer then represses transcription by tethering SIN3B to DNA. Also forms a complex with FOXK1 which represses transcription.Specifications
SIN3B | |
Polyclonal | |
Unconjugated | |
SIN3B | |
2810430C10Rik; Histone deacetylase complex subunit Sin3b; KIAA0700; Paired amphipathic helix protein Sin3b; SIN3 homolog B, transcription regulator; SIN3 homolog B, transcriptional regulator; SIN3 transcription regulator family member B; SIN3 transcription regulator homolog B; SIN3B; Transcriptional corepressor Sin3b; transcriptional regulator, SIN3 yeast homolog B; transcriptional regulator, SIN3B; transcriptional regulator, SIN3B (yeast) | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20467, 23309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunoprecipitation, Western Blot | |
0.57 mg/mL | |
0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
O75182, Q62141 | |
SIN3B | |
Recombinant fragment corresponding to a region within amino acids 15 and 245 of Human SIN3B | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |