Learn More
SETDB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA529101
Code : PN
Additional Details : Weight : 0.01000kg
Description
Recommended positive controls: A549, HeLa, HepG2, HCT116. Predicted reactivity: Mouse (83%), Rat (84%), Bovine (94%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
This gene encodes a histone methyltransferase. The encoded enzyme catalyzes the reaction of S-adenosyl-L-methionine and histone L-lysine to produce S-adenosyl-L-homocysteine and histone N(6)-methyl-L-lysine. The encoded protein likely functions in transcriptional repression. Alternatively spliced transcript variants have been described.Specifications
SETDB1 | |
Polyclonal | |
Unconjugated | |
SETDB1 | |
AU022152; ERG-associated protein with a SET domain, ESET; ERG-associated protein with SET domain; ESET; H3-K9-HMTase 4; H3-K9-HMTase4; histone H3-K9 methyltransferase 4; Histone-lysine N-methyltransferase SETDB1; histone-lysine N-methyltransferase, H3lysine-9 specific 4; KG1T; KIAA0067; KMT1E; Lysine N-methyltransferase 1E; mKIAA0067; SET domain bifurcated 1; SET domain ERG-associated histone methyltransferase; SET domain, bifurcated 1; SETDB1; TDRD21; tudor domain containing 21 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
9869 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
ChIP Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
1 mg/mL | |
PBS with 20% glycerol and 0.025% ProClin 300; pH 7 | |
Q15047 | |
SETDB1 | |
Recombinant fragment corresponding to a region within amino acids 421 and 700 of Human SETDB1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |