Learn More
SEMA4A, Rabbit, Purified MaxPab™ Polyclonal Antibody, Abnova™
Rabbit polyclonal antibody raised against a full-length human SEMA4A protein.
Brand: Abnova H00064218-D01P.100ug
Additional Details : Weight : 0.00010kg
Description
SEMA4A is a member of the semaphorin family of soluble and transmembrane proteins. Semaphorins are involved in guidance of axonal migration during neuronal development and in immune responses.[supplied by OMIM
Sequence: MALPALGLDPWSLLGLFLFQLLQLLLPTTTAGGGGQGPMPRVRYYAGDERRALSFFHQKGLQDFDTLLLSGDGNTLYVGAREAILALDIQDPGVPRLKNMIPWPASDRKKSECAFKKKSNETQCFNFIRVLVSYNVTHLYTCGTFAFSPACTFIELQDSYLLPISEDKVMEGKGQSPFDPAHKHTAVLVDGMLYSGTMNNFLGSEPILMRTLGSQPVLKTDNFLRWLHHDASFVAAIPSTQVVYFFFEETASEFDFFERLHTSRVARVCKNDVGGEKLLQKKWTTFLKAQLLCTQPGQLPFNVIRHAVLLPADSPTAPHIYAVFTSQWQVGGTRSSAVCAFSLLDIERVFKGKYKELNKETSRWTTYRGPETNPRPGSCSVGPSSDKALTFMKDHFLMDEQVVGTPLLVKSGVEYTRLAVETAQGLDGHSHLVMYLGTTTGSLHKAVVSGDSSAHLVEEIQLFPDPEPVRNLQLAPTQGAVFVGFSGGVWRVPRANCSVYESCVDCVLARDPHCAWDPESRTCCLLSAPNLNSWKQDMERGNPEWACASGPMSRSLRPQSRPQIIKEVLAVPNSILELPCPHLSALASYYWSHGPAAVPEASSTVYNGSLLLIVQDGVGGLYQCWATENGFSYPVISYWVDSQDQTLALDPELAGIPREHVKVPLTRVSGGAALAAQQSYWPHFVTVTVLFALVLSGALIILVASPLRALRARGKVQGCETLRPGEKAPLSREQHLQSPKECRTSASDVDADNNCLGTEVASpecifications
SEMA4A | |
Polyclonal | |
Rabbit polyclonal antibody raised against a full-length human SEMA4A protein. | |
SEMA4A | |
CORD10/FLJ12287/RP35/SEMAB/SEMB | |
Rabbit | |
Affinity chromatography | |
RUO | |
64218 | |
Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
Purified |
Immunoprecipitation, Western Blot | |
Unconjugated | |
PBS with no preservative; pH 7.4 | |
NM_022367 | |
SEMA4A | |
SEMA4A (NP_071762.2, 1 a.a. ∼ 761 a.a) full-length human protein. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |