missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-MECP2 (Ser80) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA513024
Code : LE
Additional Details : Weight : 0.01000kg
Description
This antibody is predicted to react with mouse, non-human primate and rat based on sequence homology.
MECP2 belongs to a family of nuclear proteins (including MBD1, MBD2, MBD3 and MBD4) that have a methyl-CpG binding domain. MECP2 is capable of binding specifically to methylated DNA. DNA methylation is the major modification of eukaryotic genomes and plays an essential role in mammalian development. MECP2 can also repress transcription from methylated gene promoters. In contrast to other MBD family members, MECP2 is X-linked and subject to X inactivation. MECP2 is dispensible in stem cells, but is essential for embryonic development. MECP2 gene mutations are the cause of most cases of Rett syndrome, a progressive neurologic developmental disorder and one of the most common causes of mental retardation in females.Specifications
Phospho-MECP2 (Ser80) | |
Polyclonal | |
Unconjugated | |
Mecp2 | |
1500041B07Rik; AUTSX3; D630021H01Rik; DKFZp686A24160; Mbd5; MECP2; MeCp-2 protein; methyl CpG binding protein 2; methyl-CpG binding protein 2; methyl-CpG-binding protein 2; MRX16; MRX79; MRXS13; MRXSL; PPMX; RS; RTS; RTT; testis tissue sperm-binding protein Li 41a; WBP10 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
4204 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Dot Blot | |
0.5 mg/mL | |
PBS with 0.09% sodium azide | |
P51608 | |
Mecp2 | |
KLH conjugated synthetic phosphopeptide corresponding to amino acid residues surrounding S80 of human MeCP2 | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |