missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-HSP20 (Ser16) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA538278
Code : NEW
Additional Details : Weight : 0.00900kg
Description
HSPB6 is associated with actin (see MIM 102540) and modulates smooth muscle relaxation.Specifications
Phospho-HSP20 (Ser16) | |
Polyclonal | |
Unconjugated | |
HSPB6 | |
Hsp20, heat shock 20 kDa-like protein p20, heat shock protein beta-6, HSPB6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
126393, 192245, 243912 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
O14558, Q5EBG6, P97541 | |
HSPB6 | |
A synthetic phosphopeptide derived from human HSP20 around the phosphorylation site of Ser16 (R-A-SP-A-P) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |