missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Phospho-Estrogen Receptor alpha (Ser102) Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA539788
Code : NEW
Additional Details : Weight : 0.00900kg
Description
Estrogen Receptors (ER) are members of the steroid/thyroid hormone receptor superfamily of nuclear receptors. The estrogen receptor is a ligand-activated transcription factor, that when bound to estrogen hormone, induces a conformational change that allows dimerization and binding to estrogen response elements (ERE) in DNA. When bound to EREs, ER can positively or negatively regulate gene transcription through the recruitment of coactivator or corepressor proteins. There are two different forms of the estrogen receptor, alpha and beta, encoded by separate genes (ESR1 and ESR2, respectively). Due to alternative RNA splicing, at least 4 estrogen receptor-alpha isoforms are known to exist (Isoform 1 (66 kDa), Isoform 2 (53 kDa), Isoform 3 (47 kDa), Isoform 4 (35 kDa)). Estrogen receptors are widely expressed in different tissue types and are essential for sexual development and reproductive function. They also play a role in other tissues such as bone. Estrogen receptors are involved in pathological processes including breast cancer, endometrial cancer, and osteoporosis.Specifications
Phospho-Estrogen Receptor alpha (Ser102) | |
Polyclonal | |
Unconjugated | |
ESR1 | |
ER, ER-alpha, Estradiol receptor, Nuclear receptor subfamily 3 group A member 1, ESR1, ESR, NR3A1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
13982, 2099, 24890 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin) | |
1 mg/mL | |
Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 | |
P03372, P19785, P06211 | |
ESR1 | |
A synthetic phosphopeptide derived from human Estrogen Receptor-alpha around the phosphorylation site of Ser102 (L-N-SP-V-S) | |
100 μg | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |